Learn More
Grainger Johns Manville 90° Elbow, Mineral Wool, 1 1/2 in. Thick, 3 in. ID
Catalog No. 502778876
Encompass_Preferred
This mineral wool 90° elbow pipe insulation is designed for industrial and commercial thermal control and fire-resistance applications. Composed of inorganic basalt fibers and a thermosetting resin, it provides high-temperature performance up to 1200°F (650°C). This unslit, preformed insulation is engineered for 2 1/2 in. iron pipe sizes with a 3 in. inner diameter and 1 1/2 in. wall thickness.
  • Water-repellent construction tested to BS EN 13472
  • Lightweight, low-dust design for clean handling and easy fabrication with a knife
  • Excellent acoustical performance meeting ISO 15665 classes A3, B3, C3, and D3
  • Non-combustible per ISO 1182
  • Resistant to mold and fungi growth
  • Low in-service shrinkage
Catalog No. 50-277-8876 Supplier Grainger Supplier No. 19NH49
This mineral wool 90° elbow pipe insulation is designed for industrial and commercial thermal control and fire-resistance applications. Composed of inorganic basalt fibers and a thermosetting resin, it provides high-temperature performance up to 1200°F (650°C). This unslit, preformed insulation is engineered for 2 1/2 in. iron pipe sizes with a 3 in. inner diameter and 1 1/2 in. wall thickness.
  • Water-repellent construction tested to BS EN 13472
  • Lightweight, low-dust design for clean handling and easy fabrication with a knife
  • Excellent acoustical performance meeting ISO 15665 classes A3, B3, C3, and D3
  • Non-combustible per ISO 1182
  • Resistant to mold and fungi growth
  • Low in-service shrinkage