Learn More
Grainger Johns Manville Pipe Fitting Insulation, Mineral Wool, 90° Elbow, 3 in. ID, 1 in. Thick, 1 ea
Catalog No. 502778871
Encompass_Preferred
This mineral wool 90° elbow pipe fitting insulation provides reliable thermal performance for commercial and industrial piping systems. Designed for use on pipe elbows, this rigid, unfaced insulation offers excellent dimensional stability across a wide temperature range of 0°F to 1,200°F. It helps minimize heat loss, contributing to energy efficiency and controlled operating costs in systems including high-pressure steam, hot water, and chilled water lines.
  • High dimensional stability and low shrinkage characteristics help reduce the potential for gaps forming at joints
  • Rigid design with water-repellent properties
  • Non-combustible insulation material suitable for high-temperature applications up to 1,200°F
  • Features an inside diameter of 3 in and a wall thickness of 1 in
  • Plenum rated for versatile installation
Catalog No. 50-277-8871 Supplier Grainger Supplier No. 19NH38
This mineral wool 90° elbow pipe fitting insulation provides reliable thermal performance for commercial and industrial piping systems. Designed for use on pipe elbows, this rigid, unfaced insulation offers excellent dimensional stability across a wide temperature range of 0°F to 1,200°F. It helps minimize heat loss, contributing to energy efficiency and controlled operating costs in systems including high-pressure steam, hot water, and chilled water lines.
  • High dimensional stability and low shrinkage characteristics help reduce the potential for gaps forming at joints
  • Rigid design with water-repellent properties
  • Non-combustible insulation material suitable for high-temperature applications up to 1,200°F
  • Features an inside diameter of 3 in and a wall thickness of 1 in
  • Plenum rated for versatile installation