Learn More
Grainger FOAMGLAS Pipe Fitting Insulation, 90° Elbow, Closed Cell Glass, 1 in. Thick, 1 ea.
Catalog No. 502783026
Encompass_Preferred
This cellular glass insulation is specifically engineered for 90° elbow pipe fittings, providing a rigid and durable solution for diverse thermal management needs. Designed for rapid field application, the insulation is factory-trimmed and preformed into two halves to facilitate straightforward assembly on piping systems. Its closed-cell structure ensures high moisture and corrosion resistance, making it suitable for demanding environments including cryogenic and high-temperature applications.
  • Operating temperature range from -450°F to +900°F
  • Rigid design for consistent thermal protection
  • Resistant to microbes, mold, and corrosive environments
  • Non-combustible material composition
  • Facilitates quick installation with split, preformed halves
Catalog No. 50-278-3026 Supplier Grainger Supplier No. 19NG64
This cellular glass insulation is specifically engineered for 90° elbow pipe fittings, providing a rigid and durable solution for diverse thermal management needs. Designed for rapid field application, the insulation is factory-trimmed and preformed into two halves to facilitate straightforward assembly on piping systems. Its closed-cell structure ensures high moisture and corrosion resistance, making it suitable for demanding environments including cryogenic and high-temperature applications.
  • Operating temperature range from -450°F to +900°F
  • Rigid design for consistent thermal protection
  • Resistant to microbes, mold, and corrosive environments
  • Non-combustible material composition
  • Facilitates quick installation with split, preformed halves