Learn More
Grainger Johns Manville Pipe Fitting Insulation, Mineral Wool, 90° Elbow, 1 1/2 in. Thick, 1 1/8 in. ID, 1 ea.
Catalog No. 502778892
Encompass_Preferred
This mineral wool insulation is designed for 90° elbow fittings in mechanical and power systems. It is factory preformed and cut-to-length to improve installation efficiency by saving fabrication time in the field.
  • Provides high dimensional stability and favorable thermal conductivity to help minimize heat loss.
  • Features low shrinkage characteristics to reduce the potential for gaps forming at the joints.
  • Rated for a temperature range of 0°F to 1,200°F.
  • Plenum rated for diverse application environments.
  • Designed for 3/4 in. nominal pipe size with 1-1/2 in. wall thickness.
  • Complies with various industry standards including ASTM C547, ASTM E84, and US Coast Guard 164.155.
Catalog No. 50-277-8892 Supplier Grainger Supplier No. 19NH44
This mineral wool insulation is designed for 90° elbow fittings in mechanical and power systems. It is factory preformed and cut-to-length to improve installation efficiency by saving fabrication time in the field.
  • Provides high dimensional stability and favorable thermal conductivity to help minimize heat loss.
  • Features low shrinkage characteristics to reduce the potential for gaps forming at the joints.
  • Rated for a temperature range of 0°F to 1,200°F.
  • Plenum rated for diverse application environments.
  • Designed for 3/4 in. nominal pipe size with 1-1/2 in. wall thickness.
  • Complies with various industry standards including ASTM C547, ASTM E84, and US Coast Guard 164.155.