Learn More
Grainger FOAMGLAS 90° Elbow Pipe Fitting Insulation, Cellular Glass, 1-3/8 in. ID, 1 in. Thick, 1 ea
Catalog No. 502778977
Encompass_Preferred
This 90° elbow pipe fitting insulation is a lightweight, rigid solution composed of millions of completely sealed cellular glass cells. Designed for industrial and commercial applications, this insulation provides consistent thermal performance for piping systems carrying water, steam, gas, and petrochemicals. The inorganic, non-combustible material is resistant to moisture, corrosion, and vermin, ensuring long-term structural and thermal stability across a wide service temperature range.
  • Constant insulating efficiency
  • Non-combustible
  • Corrosion-resistant
  • Long-term dimensional stability
  • Vermin-resistant
  • High compressive strength
  • Ecologically friendly and sustainable
  • Does not absorb water or lose effectiveness over time
Catalog No. 50-277-8977 Supplier Grainger Supplier No. 19NG65
This 90° elbow pipe fitting insulation is a lightweight, rigid solution composed of millions of completely sealed cellular glass cells. Designed for industrial and commercial applications, this insulation provides consistent thermal performance for piping systems carrying water, steam, gas, and petrochemicals. The inorganic, non-combustible material is resistant to moisture, corrosion, and vermin, ensuring long-term structural and thermal stability across a wide service temperature range.
  • Constant insulating efficiency
  • Non-combustible
  • Corrosion-resistant
  • Long-term dimensional stability
  • Vermin-resistant
  • High compressive strength
  • Ecologically friendly and sustainable
  • Does not absorb water or lose effectiveness over time