Learn More
Grainger FOAMGLAS Pipe Fitting Insulation, 90° Elbow, Cellular Glass, 1 1/2 in. Thick, 1 3/8 in. ID, 1 ea.
Catalog No. 502778978
Encompass_Preferred
This preformed, rigid cellular glass insulation is designed for 90° elbow pipe fittings. Factory-trimmed to save field fabrication time, the insulation is supplied in halves and requires sealing and banding for installation. The material is composed of inorganic, closed-cell soda lime glass, offering robust protection in demanding environments.
  • Operating temperature range: -450°F to +900°F (-268°C to +482°C)
  • R-value of 5.2 with constant insulating efficiency
  • Noncombustible and plenum rated with a flame spread index of 0
  • Excellent dimensional stability; does not shrink or swell
  • Impervious to common acids and resistant to moisture, mold, and microbes
  • Acceptable for use with stainless steel
Catalog No. 50-277-8978 Supplier Grainger Supplier No. 19NG76
This preformed, rigid cellular glass insulation is designed for 90° elbow pipe fittings. Factory-trimmed to save field fabrication time, the insulation is supplied in halves and requires sealing and banding for installation. The material is composed of inorganic, closed-cell soda lime glass, offering robust protection in demanding environments.
  • Operating temperature range: -450°F to +900°F (-268°C to +482°C)
  • R-value of 5.2 with constant insulating efficiency
  • Noncombustible and plenum rated with a flame spread index of 0
  • Excellent dimensional stability; does not shrink or swell
  • Impervious to common acids and resistant to moisture, mold, and microbes
  • Acceptable for use with stainless steel