Learn More
Grainger FOAMGLAS Pipe Fitting Insulation, 90° Elbow, Closed Cell Glass, 2 in. Thick, 1 3/8 in. ID, 6.9 R-Value
Catalog No. 502778809
Encompass_Preferred
This cellular glass insulation is designed for 90° elbow pipe fittings, providing a rigid, high-performance thermal barrier. Factory-trimmed and preformed for efficient field installation, this black, unfaced insulation is suitable for a variety of piping systems, including cryogenic, chilled water, and hydrocarbon processing applications. The closed-cell glass material offers robust resistance to moisture, mold, and corrosive environments.
  • Fits 1 in. iron pipe size with 1 3/8 in. inside diameter
  • Maintains an R-value of 6.9
  • Operates across a temperature range of -450°F to +900°F
  • Features rigid, two-piece design for assembly
  • Plenum rated with a flame spread index of 25
  • Provides resistance to microbes, mold, mildew, and corrosion
Catalog No. 50-277-8809 Supplier Grainger Supplier No. 19NG87
This cellular glass insulation is designed for 90° elbow pipe fittings, providing a rigid, high-performance thermal barrier. Factory-trimmed and preformed for efficient field installation, this black, unfaced insulation is suitable for a variety of piping systems, including cryogenic, chilled water, and hydrocarbon processing applications. The closed-cell glass material offers robust resistance to moisture, mold, and corrosive environments.
  • Fits 1 in. iron pipe size with 1 3/8 in. inside diameter
  • Maintains an R-value of 6.9
  • Operates across a temperature range of -450°F to +900°F
  • Features rigid, two-piece design for assembly
  • Plenum rated with a flame spread index of 25
  • Provides resistance to microbes, mold, mildew, and corrosion