Learn More
Grainger FOAMGLAS Pipe Fitting Insulation, 90° Elbow, Cellular Glass, 1.5 in. Thick, 1.625 in. ID, 5.2 R-Value, 1 ea.
Catalog No. 502780134
Encompass_Preferred
This cellular glass insulation is designed for 90° pipe elbow fittings. The rigid, closed-cell construction provides effective thermal protection across a wide temperature range and is engineered for use in corrosive environments.
  • Factory trimmed and preformed for efficient installation
  • Suitable for temperatures from -450°F to 900°F
  • Features 1 1/2 in. wall thickness with an R-value of 5.2
  • Resists mold, microbes, insects, and small animals
  • Provides moisture and corrosion resistance
  • Plenum rated and non-combustible
Catalog No. 50-278-0134 Supplier Grainger Supplier No. 19NG77
This cellular glass insulation is designed for 90° pipe elbow fittings. The rigid, closed-cell construction provides effective thermal protection across a wide temperature range and is engineered for use in corrosive environments.
  • Factory trimmed and preformed for efficient installation
  • Suitable for temperatures from -450°F to 900°F
  • Features 1 1/2 in. wall thickness with an R-value of 5.2
  • Resists mold, microbes, insects, and small animals
  • Provides moisture and corrosion resistance
  • Plenum rated and non-combustible