Learn More
Grainger FOAMGLAS Pipe Fitting Insulation, 90° Elbow, Cellular Glass, 2 in. Thick, 1 5/8 in. ID
Catalog No. 502778982
Encompass_Preferred
This cellular glass insulation is designed for 90° elbow pipe fittings, providing a rigid and efficient thermal solution. Factory-trimmed and preformed in halves, these components are engineered to save installation time in the field. The closed-cell glass construction offers robust performance across a wide temperature range, making it suitable for demanding industrial piping systems.
  • Provides a thermal R-value of 6.9
  • Features a nominal inside diameter of 1-5/8 in. and a wall thickness of 2 in.
  • Constructed from rigid, closed-cell glass for enhanced durability
  • Operates effectively in temperatures from -450°F to +900°F
  • Resists corrosion, moisture, mold, mildew, and microbial growth
  • Plenum rated with a flame spread index of 25
  • Meets ASTM E84, E136, and E96 standards
Catalog No. 50-277-8982 Supplier Grainger Supplier No. 19NG88
This cellular glass insulation is designed for 90° elbow pipe fittings, providing a rigid and efficient thermal solution. Factory-trimmed and preformed in halves, these components are engineered to save installation time in the field. The closed-cell glass construction offers robust performance across a wide temperature range, making it suitable for demanding industrial piping systems.
  • Provides a thermal R-value of 6.9
  • Features a nominal inside diameter of 1-5/8 in. and a wall thickness of 2 in.
  • Constructed from rigid, closed-cell glass for enhanced durability
  • Operates effectively in temperatures from -450°F to +900°F
  • Resists corrosion, moisture, mold, mildew, and microbial growth
  • Plenum rated with a flame spread index of 25
  • Meets ASTM E84, E136, and E96 standards