Learn More
Grainger Johns Manville Mineral Wool Pipe Fitting Insulation, 90° Elbow, 1-1/2 in. Thick, 10-3/4 in. ID, 1 ea
Catalog No. 502778891
Encompass_Preferred
This mineral wool 90° elbow pipe fitting insulation provides high-temperature thermal control and fire resistance for 10 in. pipes. Designed for diverse mechanical and power systems, this rigid, unfaced insulation is manufactured from mineral wool and rated for temperatures up to 1,200°F (649°C). Its high dimensional stability helps maintain effective thermal performance and minimize heat loss in demanding applications.
  • Plenum rated for safety in air handling spaces
  • Flame spread index of 25
  • Service temperature range of 0°F to 1,200°F
  • R-value of 6.5 for thermal efficiency
  • Unslit installation style
  • Fits 10 in. nominal pipe size with a 10 3/4 in. inside diameter and 1 1/2 in. wall thickness
Catalog No. 50-277-8891 Supplier Grainger Supplier No. 19NH55
This mineral wool 90° elbow pipe fitting insulation provides high-temperature thermal control and fire resistance for 10 in. pipes. Designed for diverse mechanical and power systems, this rigid, unfaced insulation is manufactured from mineral wool and rated for temperatures up to 1,200°F (649°C). Its high dimensional stability helps maintain effective thermal performance and minimize heat loss in demanding applications.
  • Plenum rated for safety in air handling spaces
  • Flame spread index of 25
  • Service temperature range of 0°F to 1,200°F
  • R-value of 6.5 for thermal efficiency
  • Unslit installation style
  • Fits 10 in. nominal pipe size with a 10 3/4 in. inside diameter and 1 1/2 in. wall thickness