Learn More
Grainger FOAMGLAS Pipe Fitting Insulation, 90° Elbow, Cellular Glass, 1 1/2 in. Thick, 1 ea.
Catalog No. 502783027
Encompass_Preferred
This 90° elbow pipe fitting insulation is constructed from rigid, closed-cell cellular glass designed for demanding thermal applications. It provides high-performance insulation across an extreme temperature range of -450°F to +900°F, making it suitable for chemical, cryogenic, and hot oil piping systems.
  • Resists moisture, mold, mildew, and corrosion
  • Features a 5.2 R-value for efficient thermal retention
  • Designed for 2 in. nominal pipe size with a 2 3/8 in. inside diameter
  • Plenum rated and non-combustible
  • Rigid closed-cell glass structure resists small insects and animals
Catalog No. 50-278-3027 Supplier Grainger Supplier No. 19NG79
This 90° elbow pipe fitting insulation is constructed from rigid, closed-cell cellular glass designed for demanding thermal applications. It provides high-performance insulation across an extreme temperature range of -450°F to +900°F, making it suitable for chemical, cryogenic, and hot oil piping systems.
  • Resists moisture, mold, mildew, and corrosion
  • Features a 5.2 R-value for efficient thermal retention
  • Designed for 2 in. nominal pipe size with a 2 3/8 in. inside diameter
  • Plenum rated and non-combustible
  • Rigid closed-cell glass structure resists small insects and animals