Learn More
Grainger FOAMGLAS Pipe Fitting Insulation, 90° Elbow, Cellular Glass, 1 in Thick, 3 1/2 in ID, 1 ea
Catalog No. 502783538
Encompass_Preferred
This cellular glass insulation is designed for 90° elbow fittings, offering a rigid, factory-trimmed solution for industrial piping systems. It is engineered for applications ranging from cryogenic systems to hot oil transport, providing high thermal efficiency and protection in corrosive environments. The insulation is supplied in halves, allowing for secure sealing and banding during installation.
  • Provides a temperature operating range of -450°F to +900°F (-270°C to 480°C)
  • Features a closed-cell structure that is moisture-resistant and corrosion-resistant
  • Resists mold, mildew, microbes, and small insects or animals
  • Offers an R-value of 3.5 for effective thermal management
  • Plenum rated with an ASTM E84 flame spread index of 25
  • Designed for use with 3 in. nominal iron pipe size fittings
Catalog No. 50-278-3538 Supplier Grainger Supplier No. 19NG70
This cellular glass insulation is designed for 90° elbow fittings, offering a rigid, factory-trimmed solution for industrial piping systems. It is engineered for applications ranging from cryogenic systems to hot oil transport, providing high thermal efficiency and protection in corrosive environments. The insulation is supplied in halves, allowing for secure sealing and banding during installation.
  • Provides a temperature operating range of -450°F to +900°F (-270°C to 480°C)
  • Features a closed-cell structure that is moisture-resistant and corrosion-resistant
  • Resists mold, mildew, microbes, and small insects or animals
  • Offers an R-value of 3.5 for effective thermal management
  • Plenum rated with an ASTM E84 flame spread index of 25
  • Designed for use with 3 in. nominal iron pipe size fittings