Learn More
Grainger FOAMGLAS Pipe Fitting Insulation, 90° Elbow, Cellular Glass, 1 1/2 in Thick, 4 1/2 in ID, 1 ea
Catalog No. 502782732
Encompass_Preferred
This cellular glass insulation is designed for 90° elbow fittings, offering a rigid, high-performance solution for demanding industrial environments. It is factory-trimmed and preformed to reduce on-site fabrication time, requiring sealing and banding of the two halves during installation.
  • Designed for pipe sizes of 4 in. with an inner diameter of 4-1/2 in. and a wall thickness of 1-1/2 in.
  • Constructed from durable, closed-cell glass material.
  • Operating temperature range of -450°F to +900°F.
  • Resistant to corrosion, moisture, mold, mildew, and pests.
  • Suitable for environments including chilled water, cryogenic systems, hot oil, and hydrocarbon processing.
  • Plenum rated with an R-value of 5.2.
  • Flame spread index of 25.
  • Complies with ASTM E84, E136, and E96 standards.
Catalog No. 50-278-2732 Supplier Grainger Supplier No. 19NG82
This cellular glass insulation is designed for 90° elbow fittings, offering a rigid, high-performance solution for demanding industrial environments. It is factory-trimmed and preformed to reduce on-site fabrication time, requiring sealing and banding of the two halves during installation.
  • Designed for pipe sizes of 4 in. with an inner diameter of 4-1/2 in. and a wall thickness of 1-1/2 in.
  • Constructed from durable, closed-cell glass material.
  • Operating temperature range of -450°F to +900°F.
  • Resistant to corrosion, moisture, mold, mildew, and pests.
  • Suitable for environments including chilled water, cryogenic systems, hot oil, and hydrocarbon processing.
  • Plenum rated with an R-value of 5.2.
  • Flame spread index of 25.
  • Complies with ASTM E84, E136, and E96 standards.