Learn More
Grainger Johns Manville Mineral Wool Pipe Fitting Insulation, 90° Elbow, 4 in Pipe Size, 1 1/2 in Thick
Catalog No. 502778877
Encompass_Preferred
This water-repellent, high-temperature mineral wool pipe insulation is designed for superior thermal and acoustical control in industrial applications. Engineered from inorganic basalt fibers and a thermosetting resin, it offers excellent dimensional stability and low shrinkage, ensuring optimal performance and energy efficiency in systems such as high-pressure steam, hot water, and chilled water lines.
  • Water-repellent construction tested to BS EN 13472
  • Suitable for service temperatures up to 1,200 °F
  • Excellent acoustical performance meeting ISO 15665 standards
  • Non-combustible material according to ISO 1182
  • Lightweight and low-dust properties for easier handling
  • Zero flame spread and smoke developed ratings per ASTM E84 and UL 723
Catalog No. 50-277-8877 Supplier Grainger Supplier No. 19NH51
This water-repellent, high-temperature mineral wool pipe insulation is designed for superior thermal and acoustical control in industrial applications. Engineered from inorganic basalt fibers and a thermosetting resin, it offers excellent dimensional stability and low shrinkage, ensuring optimal performance and energy efficiency in systems such as high-pressure steam, hot water, and chilled water lines.
  • Water-repellent construction tested to BS EN 13472
  • Suitable for service temperatures up to 1,200 °F
  • Excellent acoustical performance meeting ISO 15665 standards
  • Non-combustible material according to ISO 1182
  • Lightweight and low-dust properties for easier handling
  • Zero flame spread and smoke developed ratings per ASTM E84 and UL 723