Learn More
Grainger Johns Manville 90° Elbow Pipe Fitting Insulation, Mineral Wool, 2 in Thick, 5 1/2 in ID, 1 ea
Catalog No. 502782346
Encompass_Preferred
This water-repellent mineral wool insulation is engineered for 90° elbow pipe fittings, providing effective thermal control and fire resistance for commercial HVAC applications. Composed of inorganic basalt-derived fibers bonded with a thermosetting resin, the rigid, unslit design offers high dimensional stability to minimize heat loss and prevent joint gaps. It is non-combustible and suitable for use in air-handling spaces.
  • Service temperature range from 0°F to 1,200°F
  • Excellent thermal conductivity minimizes energy loss
  • Flame spread rating of 0 and smoke developed rating of 0 per ASTM E84 and UL 723
  • Water-repellent construction tested to BS EN 13472
  • High dimensional stability with low shrinkage
  • Easy to fabricate and install on fittings
  • Plenum rated for diverse system requirements
Catalog No. 50-278-2346 Supplier Grainger Supplier No. 19NH62
This water-repellent mineral wool insulation is engineered for 90° elbow pipe fittings, providing effective thermal control and fire resistance for commercial HVAC applications. Composed of inorganic basalt-derived fibers bonded with a thermosetting resin, the rigid, unslit design offers high dimensional stability to minimize heat loss and prevent joint gaps. It is non-combustible and suitable for use in air-handling spaces.
  • Service temperature range from 0°F to 1,200°F
  • Excellent thermal conductivity minimizes energy loss
  • Flame spread rating of 0 and smoke developed rating of 0 per ASTM E84 and UL 723
  • Water-repellent construction tested to BS EN 13472
  • High dimensional stability with low shrinkage
  • Easy to fabricate and install on fittings
  • Plenum rated for diverse system requirements