Learn More
Grainger Johns Manville Mineral Wool 90° Elbow Insulation, Unslit, 1 in Thick, 5 1/2 in ID, 1 ea
Catalog No. 502782536
Encompass_Preferred
This mineral wool 90° elbow pipe insulation is a mandrel-wound, water-repellent solution designed for high-temperature thermal control and fire resistance. Composed of inorganic fibers derived from basalt and bound with a thermosetting resin, this rigid insulation provides excellent dimensional stability and performance in demanding industrial applications up to 1,200°F.
  • Water-repellent construction tested to BS EN 13472
  • Non-combustible and plenum rated
  • Low smoke and flame spread index of 25 per ASTM E84
  • Excellent acoustical performance exceeding ISO 15665 criteria
  • Lightweight and easy to cut for simplified installation
  • Mold resistant to prevent fungal growth
  • Uniform thermal conductivity of 0.23 Btu•in/(hr•ft²•°F)
Catalog No. 50-278-2536 Supplier Grainger Supplier No. 19NH41
This mineral wool 90° elbow pipe insulation is a mandrel-wound, water-repellent solution designed for high-temperature thermal control and fire resistance. Composed of inorganic fibers derived from basalt and bound with a thermosetting resin, this rigid insulation provides excellent dimensional stability and performance in demanding industrial applications up to 1,200°F.
  • Water-repellent construction tested to BS EN 13472
  • Non-combustible and plenum rated
  • Low smoke and flame spread index of 25 per ASTM E84
  • Excellent acoustical performance exceeding ISO 15665 criteria
  • Lightweight and easy to cut for simplified installation
  • Mold resistant to prevent fungal growth
  • Uniform thermal conductivity of 0.23 Btu•in/(hr•ft²•°F)