Learn More
Grainger FOAMGLAS Pipe Fitting Insulation, 90° Elbow, Cellular Glass, 2 in. Thick, 1 ea.
Catalog No. 502778972
Encompass_Preferred
This 90° elbow pipe fitting insulation is designed for use on pipe fittings in industrial and commercial applications, including cryogenic systems and hydrocarbon processing. It is constructed from rigid, closed-cell cellular glass, providing a durable and efficient thermal barrier.
  • Closed-cell cellular glass composition is non-combustible and impermeable to water and water vapor.
  • Maintains a constant insulating efficiency across a wide service temperature range of -450°F to 900°F.
  • Features long-term dimensional stability and high compressive strength.
  • Resists corrosion, chemical exposure, mold, mildew, and small insects or animals.
  • Standardized for use with stainless steel components.
  • Plenum rated with a flame spread index of 25.
Catalog No. 50-277-8972 Supplier Grainger Supplier No. 19NG93
This 90° elbow pipe fitting insulation is designed for use on pipe fittings in industrial and commercial applications, including cryogenic systems and hydrocarbon processing. It is constructed from rigid, closed-cell cellular glass, providing a durable and efficient thermal barrier.
  • Closed-cell cellular glass composition is non-combustible and impermeable to water and water vapor.
  • Maintains a constant insulating efficiency across a wide service temperature range of -450°F to 900°F.
  • Features long-term dimensional stability and high compressive strength.
  • Resists corrosion, chemical exposure, mold, mildew, and small insects or animals.
  • Standardized for use with stainless steel components.
  • Plenum rated with a flame spread index of 25.