Learn More
Grainger Johns Manville Pipe Fitting Insulation, 90° Elbow, Mineral Wool, 1-1/2 in. Thick
Catalog No. 502778944
Encompass_Preferred
This mineral wool 90° elbow pipe insulation is designed for high-temperature thermal and acoustical performance in mechanical, power, and process piping systems. This rigid, unfaced insulation offers service temperatures from 0°F to 1,200°F (-18°C to 650°C), providing thermal stability to minimize heat loss and reduce operating costs. Its water-repellent design mitigates moisture intrusion, and its non-combustible composition ensures safety in industrial environments.
  • Provides excellent thermal insulation for systems operating up to 1,200°F
  • Features high dimensional stability and low shrinkage to prevent gaps at joints
  • Water-repellent construction absorbs less than 0.35 kg/m² of water
  • Rated non-combustible per ISO 1182
  • Exceeds ISO 15665 criteria for acoustic classes A3, B3, C3, and D3
  • Easily fabricated and cut
Catalog No. 50-277-8944 Supplier Grainger Supplier No. 19NH43
This mineral wool 90° elbow pipe insulation is designed for high-temperature thermal and acoustical performance in mechanical, power, and process piping systems. This rigid, unfaced insulation offers service temperatures from 0°F to 1,200°F (-18°C to 650°C), providing thermal stability to minimize heat loss and reduce operating costs. Its water-repellent design mitigates moisture intrusion, and its non-combustible composition ensures safety in industrial environments.
  • Provides excellent thermal insulation for systems operating up to 1,200°F
  • Features high dimensional stability and low shrinkage to prevent gaps at joints
  • Water-repellent construction absorbs less than 0.35 kg/m² of water
  • Rated non-combustible per ISO 1182
  • Exceeds ISO 15665 criteria for acoustic classes A3, B3, C3, and D3
  • Easily fabricated and cut