Learn More
Grainger Johns Manville MinWool-1200 Pipe Fitting Insulation, Mineral Wool, 6 in. Pipe Size, 1 in. Wall Thickness, 1 ea.
Catalog No. 502778873
Encompass_Preferred
MinWool-1200 pipe insulation is a water-repellent, rigid insulation designed for high-temperature thermal control and fire-resistance in commercial and industrial applications. Manufactured from inorganic basalt fibers and a thermosetting resin, this pipe fitting insulation provides stable thermal performance at temperatures ranging from 0°F to 1200°F (650°C).
  • Provides excellent thermal control and energy savings with a low thermal conductivity value.
  • Features non-combustible material composition that meets ASTM E84 and UL 723 standards with a flame spread index of 25.
  • Maintains high dimensional stability with low shrinkage to prevent gaps at joints.
  • Resists mold and fungi growth per ASTM C1338 testing.
  • Meets ISO 15665 acoustical performance criteria for classes A3, B3, C3, and D3.
  • Lightweight and easy to fabricate on-site.
Catalog No. 50-277-8873 Supplier Grainger Supplier No. 19NH42
MinWool-1200 pipe insulation is a water-repellent, rigid insulation designed for high-temperature thermal control and fire-resistance in commercial and industrial applications. Manufactured from inorganic basalt fibers and a thermosetting resin, this pipe fitting insulation provides stable thermal performance at temperatures ranging from 0°F to 1200°F (650°C).
  • Provides excellent thermal control and energy savings with a low thermal conductivity value.
  • Features non-combustible material composition that meets ASTM E84 and UL 723 standards with a flame spread index of 25.
  • Maintains high dimensional stability with low shrinkage to prevent gaps at joints.
  • Resists mold and fungi growth per ASTM C1338 testing.
  • Meets ISO 15665 acoustical performance criteria for classes A3, B3, C3, and D3.
  • Lightweight and easy to fabricate on-site.