Learn More
Grainger Johns Manville Pipe Fitting Insulation, 90° Elbow, Mineral Wool, 1 1/2 in. Thick, 6 5/8 in. ID
Catalog No. 502782323
Encompass_Preferred
This mineral wool pipe fitting insulation is designed for 90° elbows on piping systems including chilled water, hot water, and high-pressure steam. This unslit, rigid insulation offers high dimensional stability and low shrinkage to minimize heat loss and prevent gap formation at joints. It is plenum rated and suitable for diverse mechanical and power system applications.
  • Operating temperature range of 0°F to +1,200°F
  • Approximate R-value of 6.5
  • Inside diameter of 6 5/8 in. and wall thickness of 1 1/2 in.
  • Compliant with ASTM C547 Type 1 and 2, ASTM E84, ASTM C585, and UL 723
  • Suitable for use on 6 in. iron pipe size
Catalog No. 50-278-2323 Supplier Grainger Supplier No. 19NH53
This mineral wool pipe fitting insulation is designed for 90° elbows on piping systems including chilled water, hot water, and high-pressure steam. This unslit, rigid insulation offers high dimensional stability and low shrinkage to minimize heat loss and prevent gap formation at joints. It is plenum rated and suitable for diverse mechanical and power system applications.
  • Operating temperature range of 0°F to +1,200°F
  • Approximate R-value of 6.5
  • Inside diameter of 6 5/8 in. and wall thickness of 1 1/2 in.
  • Compliant with ASTM C547 Type 1 and 2, ASTM E84, ASTM C585, and UL 723
  • Suitable for use on 6 in. iron pipe size