Learn More
Grainger FOAMGLAS 90° Elbow, Closed Cell Glass, 2 in. Thick, 6 5/8 in. ID
Catalog No. 502778742
Encompass_Preferred
This 90° elbow pipe fitting insulation is designed for high-performance thermal management in industrial piping systems. Constructed from rigid, closed-cell glass, this prefabricated insulation provides reliable protection across a wide temperature range and is engineered for use in chemical, chilled water, cryogenic, and hot oil piping systems.
  • Temperature range of -270°C to +480°C (-450°F to +900°F)
  • Nominal inside diameter of 6 5/8 in. with 2 in. wall thickness
  • Rigid closed-cell glass structure is waterproof, vapor-tight, and pest-proof
  • Incombustible material with a fire classification of A1
  • Acid-resistant and high compressive strength of 1.60 N/mm
  • Factory prefabricated and pre-coated with high temperature anti-abrasive finish
  • Plenum rated for installation versatility
Catalog No. 50-277-8742 Supplier Grainger Supplier No. 19NG94
This 90° elbow pipe fitting insulation is designed for high-performance thermal management in industrial piping systems. Constructed from rigid, closed-cell glass, this prefabricated insulation provides reliable protection across a wide temperature range and is engineered for use in chemical, chilled water, cryogenic, and hot oil piping systems.
  • Temperature range of -270°C to +480°C (-450°F to +900°F)
  • Nominal inside diameter of 6 5/8 in. with 2 in. wall thickness
  • Rigid closed-cell glass structure is waterproof, vapor-tight, and pest-proof
  • Incombustible material with a fire classification of A1
  • Acid-resistant and high compressive strength of 1.60 N/mm
  • Factory prefabricated and pre-coated with high temperature anti-abrasive finish
  • Plenum rated for installation versatility