Learn More
Grainger Johns Manville Mineral Wool Pipe Fitting Insulation, 90 Degrees Elbow, 8 5/8 in. I.D., 1 1/2 in. Wall
Catalog No. 502782532
Encompass_Preferred
This mineral wool elbow pipe fitting insulation is designed for 8 in. piping systems, offering efficient thermal performance and rigid protection for 90-degree elbows. This unfaced insulation is suitable for a variety of applications, including chilled water, hot water, and high-pressure steam systems.
  • High dimensional stability and good thermal conductivity help minimize heat loss
  • Low shrinkage characteristics reduce the potential for gaps forming at the joints
  • Operating temperature range of 0°F to 1,200°F
  • Plenum rated with a flame spread index of 25
  • Meets ASTM C547 Type I and II, ASTM C585, ASTM E84, E136, NRC Reg Guide 1.36, UL 723, and US Coast Guard 164.165 standards
Catalog No. 50-278-2532 Supplier Grainger Supplier No. 19NH54
This mineral wool elbow pipe fitting insulation is designed for 8 in. piping systems, offering efficient thermal performance and rigid protection for 90-degree elbows. This unfaced insulation is suitable for a variety of applications, including chilled water, hot water, and high-pressure steam systems.
  • High dimensional stability and good thermal conductivity help minimize heat loss
  • Low shrinkage characteristics reduce the potential for gaps forming at the joints
  • Operating temperature range of 0°F to 1,200°F
  • Plenum rated with a flame spread index of 25
  • Meets ASTM C547 Type I and II, ASTM C585, ASTM E84, E136, NRC Reg Guide 1.36, UL 723, and US Coast Guard 164.165 standards