Learn More
Grainger FOAMGLAS Pipe Fitting Insulation, 90° Elbow, Cellular Glass, 1-1/2 in. Thick, 8-5/8 in. ID
Catalog No. 502778981
Encompass_Preferred
This cellular glass pipe fitting insulation is a rigid, black, 90° elbow designed for use in industrial and commercial piping systems. Composed of millions of completely sealed glass cells, this material offers superior thermal performance and dimensional stability across a wide temperature range. It is suitable for applications including cryogenic systems, hydrocarbon processing, and chemical handling due to its non-combustible and corrosion-resistant properties.
  • Service temperature range of -450°F to +900°F
  • Impermeable to water and water vapor with zero capillarity
  • High compressive strength and long-term dimensional stability
  • Non-combustible and plenum rated
  • Resistant to corrosion, vermin, mold, and microbes
  • Approximate R-value of 5.2
  • Designed for 8 in. pipe size with 1 1/2 in. wall thickness
Catalog No. 50-277-8981 Supplier Grainger Supplier No. 19NG85
This cellular glass pipe fitting insulation is a rigid, black, 90° elbow designed for use in industrial and commercial piping systems. Composed of millions of completely sealed glass cells, this material offers superior thermal performance and dimensional stability across a wide temperature range. It is suitable for applications including cryogenic systems, hydrocarbon processing, and chemical handling due to its non-combustible and corrosion-resistant properties.
  • Service temperature range of -450°F to +900°F
  • Impermeable to water and water vapor with zero capillarity
  • High compressive strength and long-term dimensional stability
  • Non-combustible and plenum rated
  • Resistant to corrosion, vermin, mold, and microbes
  • Approximate R-value of 5.2
  • Designed for 8 in. pipe size with 1 1/2 in. wall thickness