Learn More
Cole-Parmer Idex Fingertight One-Piece Fitting, Standard Knurl, Black PEEK, 1/16" OD Tubing, 10-32 Coned; 1/EA
Catalog No. NC3737994 Shop All Cole-Parmer Products
Encompass
One-Piece Fingertight FittingsGet the convenience of a nut and ferrule built into one fitting. No need to worry the ferrule could get stuck in the receiving port! Fittings are available in various colors, materials and port sizes to suit virtually every application. Fittings with headless knurl are ideal for tight spaces, while diamond knurl provides an extra grip for tightening the fitting.Easily tighten fittings without toolsVariety of fittings to suit your needsOne-piece fittings of a nut and ferrule saves timeBlack PEEK
Catalog No. NC3737994 Supplier Cole-Parmer Supplier No. 0201877
One-Piece Fingertight FittingsGet the convenience of a nut and ferrule built into one fitting. No need to worry the ferrule could get stuck in the receiving port! Fittings are available in various colors, materials and port sizes to suit virtually every application. Fittings with headless knurl are ideal for tight spaces, while diamond knurl provides an extra grip for tightening the fitting.Easily tighten fittings without toolsVariety of fittings to suit your needsOne-piece fittings of a nut and ferrule saves timeBlack PEEK