Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) LCD24004 Metal quick-disconnect coupling insert, valved, 1/4"" NPT (M) thread, 1/4"" flow size
Catalog No. 50627786
Encompass
Metal quick-disconnect couplings require a fitting body and an insert with the same flow size (order separately). Withstand vacuum to 250 psi, temperatures from -40 to 180¿F, and can be used with gases or liquids.
Catalog No. 50-627-786 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3140350
Metal quick-disconnect couplings require a fitting body and an insert with the same flow size (order separately). Withstand vacuum to 250 psi, temperatures from -40 to 180¿F, and can be used with gases or liquids.