Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) LCD22004 Quick-disconnect fittings, inserts,valved fittings, hose barb, 1/4"" ID, 1.35 L
Catalog No. 50627783
Encompass
Metal quick-disconnect couplings require a fitting body and an insert with the same flow size (order separately). Withstand vacuum to 250 psi, temperatures from -40 to 180¿F, and can be used with gases or liquids.
Catalog No. 50-627-783 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3140340
Metal quick-disconnect couplings require a fitting body and an insert with the same flow size (order separately). Withstand vacuum to 250 psi, temperatures from -40 to 180¿F, and can be used with gases or liquids.