Learn More
Nordson Medical FITTING MINI FLANGE 3/8 100PK
Catalog No. NC2158319
Encompass_Preferred
Sanitary Fitting, Mini Flange to 600 Series Barb, 3/8" (9.5 mm) Tubing ID, Natural Kynar PVDF, 100pc/pack
This item is not returnable. View return policy
Catalog No. NC2158319 Supplier Nordson Medical Supplier No. SFMN670J2A
Sanitary Fitting, Mini Flange to 600 Series Barb, 3/8" (9.5 mm) Tubing ID, Natural Kynar PVDF, 100pc/pack