Learn More
Hach Company Hach PEEK fitting, 1/16 in tubing x 1 in length (for Lachat FIA valves)
Catalog No. NC2157420 Shop All Hach Company Products
Encompass
Hach PEEK fitting designed for 1/16 in tubing with a 1 in length, intended for analytical fluidics and flow injection analysis applications such as Lachat FIA valves.
  • Chemically resistant PEEK material.
  • Fits 1/16 in outer diameter tubing.
  • 1 in length for secure connections.
  • Designed to minimize leaks in analytical instrument fluidics.
This item is not returnable. View return policy
Catalog No. NC2157420 Supplier Hach Company Supplier No. 85245
Hach PEEK fitting designed for 1/16 in tubing with a 1 in length, intended for analytical fluidics and flow injection analysis applications such as Lachat FIA valves.
  • Chemically resistant PEEK material.
  • Fits 1/16 in outer diameter tubing.
  • 1 in length for secure connections.
  • Designed to minimize leaks in analytical instrument fluidics.