Learn More
AVANTOR MASTERFLEX (VWR LLC) FITTING RED.1/4FTFTX5/32FTFT 1
Catalog No. 502742296
Encompass
Masterflex Fitting, Polypropylene, Straight, Hose Barb Reducer, 1/4IN ID x 5/32IN ID; 10/PK, Avantor
Catalog No. 50-274-2296 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4062226
Masterflex Fitting, Polypropylene, Straight, Hose Barb Reducer, 1/4IN ID x 5/32IN ID; 10/PK, Avantor