Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, HDPE, Straight, Hose Barb Reducer, 3/4"" ID x 3/8"" ID; 50/PK
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3062607
Encompass
These economical straight barbed reducing fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
Catalog No. 50-629-347