Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Nylon, Straight, Hose Barb Reducer, 5/8"" ID x 1/2"" ID; 100/PK
Catalog No. 50629331
Encompass
These economical straight barbed reducing fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
Catalog No. 50-629-331 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3062604
These economical straight barbed reducing fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.