Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, HDPE, Straight, Hose Barb Reducer, 1/4"" ID x 1/8"" ID, pack of 10
Catalog No. 50629476
Encompass
These economical straight barbed reducing fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
Catalog No. 50-629-476 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3062227
These economical straight barbed reducing fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.