Learn More
Waters Corp Rupture Disc Housing Fitting, 1/4in Disc
Catalog No. 502390509 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Rupture Disc Housing Fitting is a component for laboratory systems, designed to provide emergency pressure relief. It helps install new rupture discs, which are crucial for replacing existing discs to ensure proper function and allow assays to continue. This component has been strictly tested for quality control.
  • Provides emergency pressure relief.
  • Helps install new rupture discs.
  • Strictly tested for quality control.
  • Prevents over-pressurization in lab equipment.
  • Ensures proper function of laboratory systems.
Catalog No. 50-239-0509 Supplier Waters Corp Supplier No. 700006538
The Rupture Disc Housing Fitting is a component for laboratory systems, designed to provide emergency pressure relief. It helps install new rupture discs, which are crucial for replacing existing discs to ensure proper function and allow assays to continue. This component has been strictly tested for quality control.
  • Provides emergency pressure relief.
  • Helps install new rupture discs.
  • Strictly tested for quality control.
  • Prevents over-pressurization in lab equipment.
  • Ensures proper function of laboratory systems.