Learn More
New Jersey Fluid Systems LLC FITTING SS-4-TA-7-4FITTING
Catalog No. NC9681696
Encompass
316 Stainless Steel Swagelok Tube Fitting Adapter. 1/4 in. OD Tube Stub x 1/4 in. Female NPT
Catalog No. NC9681696 Supplier New Jersey Fluid Systems LLC Supplier No. SS4TA74
316 Stainless Steel Swagelok Tube Fitting Adapter. 1/4 in. OD Tube Stub x 1/4 in. Female NPT