Learn More
Waters Corp Waters Fitting, Stainless Steel, 1/16 mm ID, 5 per pack
Supplier:  Waters Corp WAT025604
Encompass_Preferred
This pack contains 5 compression screws and 5 annealed ferrules. The components are precisely manufactured with specific inside and outside diameters to ensure leak-free connections. Highest cleanliness standards and practices in handling and packaging eliminate debris that could damage valves and block columns.
  • Contains 5 compression screws and 5 annealed ferrules.
  • Manufactured with precise inside and outside diameters to ensure leak-free connections.
  • Made from stainless steel.
  • Internal diameter of 1/16 mm.
  • Packaged to high cleanliness standards to eliminate debris that could damage valves and block columns.
  • Suitable for HPLC and purification systems.
Catalog No. 50-819-016