Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Nylon, Straight, Hose Barb Union, 3/16"" ID; 100/PK
Catalog No. 50629593
Encompass
These economical straight barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
Catalog No. 50-629-593 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3062656
These economical straight barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.