Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, HDPE, Straight, Hose Barb Union, 1/4"" ID; 10/PK
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3062192
Encompass
These fittings with quick and easy assembly are ideal for general purpose and single-use applications to connect tubing with the rest of the system. Determine which connector type is needed for your application by contacting technical application assistance, which is free of charge. Its single-use custom configuration solutions help to minimize downtime. The fittings are always in stock with the majority of materials and configurations available to ship the same day. Also, there are unique packaging options like individuals, packs and kits for customer-friendly ordering, as well as delivery.
Catalog No. 50-629-266