Learn More
Cole-Parmer Cole-Parmer Sanitary to Hosebarb Fitting, Straight Adapter, Polypropylene, 3/4" Mini Tri-Clamp x 1/8" ID; 1/Ea
Catalog No. NC3162590 Shop All Cole-Parmer Products
Encompass_Preferred
High-quality nonmetallic sanitary fittings for critical fluid applicationsMeet US and EU standards for use in pharmaceutical process applicationsASME-BPE, USP Class VI, and FDA compliantSterilize by autoclave, EtO, or gamma irradiationIdeal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
Catalog No. NC3162590 Supplier Cole-Parmer Supplier No. 5011901
High-quality nonmetallic sanitary fittings for critical fluid applicationsMeet US and EU standards for use in pharmaceutical process applicationsASME-BPE, USP Class VI, and FDA compliantSterilize by autoclave, EtO, or gamma irradiationIdeal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system