Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) MPX22803M Quick-disconnect fittings, Polycarbonate fittings, insert, 1/2"" ID, 5/pack
Catalog No. 50627918
Encompass
All-plastic quick-disconnect couplings require a standard fitting body and standard insert or large fitting body and large insert (order separately). Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).
Catalog No. 50-627-918 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3131163
All-plastic quick-disconnect couplings require a standard fitting body and standard insert or large fitting body and large insert (order separately). Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).