Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) Quick-disconnect Complete Coupling Fittings, Polycarbonate, 1/4"" ID; 10/pk
Catalog No. 50627531
Encompass
All-plastic quick-disconnect couplings include a fitting body and insert. Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).
Catalog No. 50-627-531 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3130500
All-plastic quick-disconnect couplings include a fitting body and insert. Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).