Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) MPC17004T03 Quick-Disconnect Body, PC, 1/4"" ID; 5/Pk
Catalog No. 50627865
Encompass
All-plastic quick-disconnect couplings require a standard fitting body and standard insert or large fitting body and large insert (order separately). Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).
Catalog No. 50-627-865 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3131111
All-plastic quick-disconnect couplings require a standard fitting body and standard insert or large fitting body and large insert (order separately). Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).