Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) MPC22006T03M Quick-disconnect fittings, Polycarbonate fittings, PP, 3/8"" ID, insert, 5/pack
Catalog No. 50627880
Encompass
All-plastic quick-disconnect couplings require a standard fitting body and standard insert or large fitting body and large insert (order separately). Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).
Catalog No. 50-627-880 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3131162
All-plastic quick-disconnect couplings require a standard fitting body and standard insert or large fitting body and large insert (order separately). Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).