Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) MPC22004T39M Quick-disconnect fittings, Polysulfone fittings, 1/4"" ID, insert, 5/pack
Catalog No. 50627877
Encompass
All-plastic quick-disconnect couplings require a standard fitting body and standard insert or large fitting body and large insert (order separately). Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).
This item is not returnable. View return policy
Catalog No. 50-627-877 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3131171
All-plastic quick-disconnect couplings require a standard fitting body and standard insert or large fitting body and large insert (order separately). Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).