Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) Quick-disconnect Complete Coupling Fittings, Polysulfone, 3/8"" ID; 10/pk
Catalog No. 50627530
Encompass
All-plastic quick-disconnect couplings include a fitting body and insert. Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).
Catalog No. 50-627-530 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3130605
All-plastic quick-disconnect couplings include a fitting body and insert. Simply press the thumb latch to disconnect fitting halves. Available in polycarbonate (PC), Acetal, and polysulfone (PSF).