Learn More
Med Vet International Blood Pressure Tube Set Flexiport, 2-BVL, 10/Bag
Catalog No. 503045571
Encompass_Preferred
Every cuff can be used as a one- or two-tube cuff thanks to patented flexiport technology. Every cuff size, from small infant through thigh cuffs. Choose from either soft or vinyl fabric to fit anyone's preference. Color-coded for fast, accurate selection of the correct cuff size. Rotatable port reduces stress to cuff tubing and port, improving patient comfort and cuff durability. meets all the latest clinical guidelines for proper fit from aami and aha.
Catalog No. 50-304-5571 Supplier Med Vet International Supplier No. 666250BG
Every cuff can be used as a one- or two-tube cuff thanks to patented flexiport technology. Every cuff size, from small infant through thigh cuffs. Choose from either soft or vinyl fabric to fit anyone's preference. Color-coded for fast, accurate selection of the correct cuff size. Rotatable port reduces stress to cuff tubing and port, improving patient comfort and cuff durability. meets all the latest clinical guidelines for proper fit from aami and aha.