Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fitusiran | 1499251-18-1 | 94.7% | 16,248.2 g/mol | 5 MG
SDP

Catalog No. 5000262168
Encompass_Preferred

Fitusiran is a research-grade small interfering RNA (siRNA) that targets antithrombin (AT) messenger RNA to reduce AT production in the liver, increasing thrombin generation. It is supplied as an oligonucleotide reagent for preclinical and in-vitro studies related to coagulation and hemophilia research and is not for human therapeutic use.

  • Targets antithrombin messenger RNA to lower antithrombin production.
  • Intended for preclinical and in vitro coagulation and hemophilia research.
  • Supplied as a white to off-white solid oligonucleotide.
  • Molecular weight 16,248.2 g/mol.
  • Purity 94.7%.
  • Recommended storage: 4°C dry; in solvent store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-62168 Supplier Medchemexpress LLC Supplier No. HY1325875MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fitusiran is a research-grade small interfering RNA (siRNA) that targets antithrombin (AT) messenger RNA to reduce AT production in the liver, increasing thrombin generation. It is supplied as an oligonucleotide reagent for preclinical and in-vitro studies related to coagulation and hemophilia research and is not for human therapeutic use.

  • Targets antithrombin messenger RNA to lower antithrombin production.
  • Intended for preclinical and in vitro coagulation and hemophilia research.
  • Supplied as a white to off-white solid oligonucleotide.
  • Molecular weight 16,248.2 g/mol.
  • Purity 94.7%.
  • Recommended storage: 4°C dry; in solvent store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.