Learn More
Medchemexpress LLC Fitusiran | 1499251-18-1 | 94.7% | C78H139N11O30 | 10MG
SDP
Catalog No. 5000261519
Encompass_Preferred
Fitusiran is a small interfering RNA (siRNA) that targets antithrombin (AT) messenger RNA in the liver to lower AT production and thereby increase thrombin generation. It is supplied as a research-grade, chemically modified oligonucleotide for preclinical and biochemical studies in coagulation and hemophilia research.
  • Targets antithrombin mRNA to reduce antithrombin levels.
  • Increases thrombin generation for coagulation studies.
  • Contains chemically modified nucleotides for enhanced stability.
  • Provided with reported purity suitable for research use.
  • Supplied in small pack sizes for preclinical and in vitro experiments.
Catalog No. 50-002-61519 Supplier Medchemexpress LLC Supplier No. HY13258710MG
Fitusiran is a small interfering RNA (siRNA) that targets antithrombin (AT) messenger RNA in the liver to lower AT production and thereby increase thrombin generation. It is supplied as a research-grade, chemically modified oligonucleotide for preclinical and biochemical studies in coagulation and hemophilia research.
  • Targets antithrombin mRNA to reduce antithrombin levels.
  • Increases thrombin generation for coagulation studies.
  • Contains chemically modified nucleotides for enhanced stability.
  • Provided with reported purity suitable for research use.
  • Supplied in small pack sizes for preclinical and in vitro experiments.