Learn More
Thermo Scientific Five anion standard solution, 100 mL
Catalog No. NC1566191 Shop All Thermo Scientific Products
Encompass
Anion standard solution for ion chromatography calibration, performance checks, and anion standards.
Catalog No. NC1566191 Supplier Thermo Scientific Supplier No. 037157
Anion standard solution for ion chromatography calibration, performance checks, and anion standards.