Learn More
Electron Microscopy Sciences Five-Slide Gripper, 6 per pack
SDP
Catalog No. 50931041
Encompass_Preferred
The Five-Slide Gripper accommodates 5 microscope slides in one staining procedure. It fits most coplin and round-open staining jars. It is made from a special material which is resistant to all chemicals and solvents which are used in staining. There is no need to remove slides for drying and it withstands drying temperatures up to 80 degree C.
Catalog No. 50-931-041 Supplier Electron Microscopy Sciences Supplier No. 7141006
The Five-Slide Gripper accommodates 5 microscope slides in one staining procedure. It fits most coplin and round-open staining jars. It is made from a special material which is resistant to all chemicals and solvents which are used in staining. There is no need to remove slides for drying and it withstands drying temperatures up to 80 degree C.