Learn More
Mettler Toledo™ LE703 Sensor for FiveGo™ Conductivity Meters
Catalog No. 01912368 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Length (Metric) Cable:
2 m
Catalog No. Length (Metric) Cable
01-912-368 2 m
1 options
Catalog No. 01-912-368 Supplier Mettler Toledo™ Supplier No. 30266974
Add to Cart
Add to Cart
IP67 waterproof, robust conductivity probe for medium to high conductivities.
  • Range: 10μS/cm to 200mS/cm
  • Plug/Connection: LTW
TRUSTED_SUSTAINABILITY
Specifications
Connector Types LTW (IP67)
Description LE Cond Electrode LE703-IP67
Type LE703 Sensor, IP67 Waterproof
For Use With (Application) For medium to high conductivities
Length (Metric) Cable 2 m